Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAPOLG Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130061G
Description
PAPOLG Polyclonal antibody specifically detects PAPOLG in Human, Mouse samples. It is validated for Western BlotSpecifications
PAPOLG | |
Polyclonal | |
Western Blot | |
EC 2.7.7.19, FLJ11805, FLJ12972, FLJ13482, FLJ14187, MGC133307, MGC133308, neo-PAP, Neo-poly(A) polymerase, nuclear poly(A) polymerase gamma, PAP2, PAPG, PAP-gamma, poly(A) polymerase gamma, Polynucleotide adenylyltransferase gamma, SRP RNA 3' adenylating enzyme/pap2, SRP RNA 3'-adenylating enzyme | |
maps to a region between residue 686 and 736 of human poly(A) polymerase gamma using the numbering given in entry NP_075045.2 | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
64895 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction