Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAPOLG Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541901
Description
Peptide sequence: SVDAIGGESM PIPTIDTSRK KRLPSKELPD SSSPVPANNI RVIKNSIRLT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
This gene overlaps with the gene for cysteineendopeptidase AUT-like 4 in a head-to-tail orientation.
Specifications
PAPOLG | |
Polyclonal | |
Unconjugated | |
PAPOLG | |
9630006B20Rik; FLJ11805; FLJ12972; FLJ13482; FLJ14187; hypothetical protein MGC63762; MGC133307; MGC133308; neo-PAP; Neo-poly(A) polymerase; nuclear poly(A) polymerase gamma; PAP2; PAPG; PAP-gamma; Papolg; Poly(A) polymerase gamma; Poly(A) polymerase gamma-like protein; polynucleotide adenylyltransferase gamma; Signal recognition particle RNA-adenylating enzyme; SRP 3'-adenylating protein; SRP RNA 3' adenylating enzyme/pap2; SRP RNA 3'-adenylating enzyme; SRP RNA-adenylating enzyme; zgc:63762 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64895 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BWT3 | |
PAPOLG | |
Synthetic peptide directed towards the middle region of human PAPOLG. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction