Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAPOLG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565148 Supplier Invitrogen™ Supplier No. PA565148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAAVIVEKPLS VPPAQGLSIP VIGAKVDSTV KTVSPPTVCT IPTVVGRNVI PRITTPHNPA QGQPHLNGMS NITKT Highest antigen sequence identity to the following orthologs - mouse 84%, rat 82%.

This gene overlaps with the gene for cysteineendopeptidase AUT-like 4 in a head-to-tail orientation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPOLG
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAPOLG
Gene Accession No. Q9BWT3
Gene Alias 9630006B20Rik; FLJ11805; FLJ12972; FLJ13482; FLJ14187; hypothetical protein MGC63762; MGC133307; MGC133308; neo-PAP; Neo-poly(A) polymerase; nuclear poly(A) polymerase gamma; PAP2; PAPG; PAP-gamma; Papolg; Poly(A) polymerase gamma; Poly(A) polymerase gamma-like protein; polynucleotide adenylyltransferase gamma; Signal recognition particle RNA-adenylating enzyme; SRP 3'-adenylating protein; SRP RNA 3' adenylating enzyme/pap2; SRP RNA 3'-adenylating enzyme; SRP RNA-adenylating enzyme; zgc:63762
Gene Symbols PAPOLG
Host Species Rabbit
Immunogen Recombinant Human PAPOLG. Recombinant protein control fragment (Product #RP-104661).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.