Learn More
Invitrogen™ PAPPA Monoclonal Antibody (10E2)

Description
MA1-22743 detects PAPP A in Human samples.
Specifications
Specifications
| Antigen | PAPPA |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 10E2 |
| Concentration | 5.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PAPPA |
| Gene Accession No. | Q13219 |
| Gene Alias | 8430414N03Rik; ASBABP2; aspecific BCL2 ARE-binding protein 2; differentially placenta 1 expressed protein; DIPLA1; IGFBP-4 ase; IGFBP-4ase; IGF-dependent IGFBP-4 protease; insulin-like growth factor-dependent IGF binding protein-4 protease; insulin-like growth factor-dependent IGF-binding protein 4 protease; PAG1; PAPA; PAPP A; PAPPA; PAPP-A; PAPPA1; Pappalysin; pappalysin 1; pappalysin-1; pregnacy-associated plasma protein A; pregnancy-associated plasma protein A; pregnancy-associated plasma protein A, pappalysin 1; RP11-45A16.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.