Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAPPA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579782
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Human Placenta Tissue, HT1080 whole cell, SKOV whole cell, 22RV1 whole cell, SW620 whole cell, MM231 whole cell. IHC: Human Placenta Tissue.
Pregnancy-associated plasma protein-A (PAPP-A, Insulin-like growth factor binding protein-4 protease, PAPP-A-proMBP complex) is a large heterottetrameric glycoprotein of approximately 500 kDa, which was first discovered in serum from pregnant women. The hetreotetramer consists of two PAPP-A subunits and two proMBP subunits. Low maternal serum levels of PAPP-A in first trimester biochemical screening is used as a marker of Down's syndrome (trisomy 21).
Specifications
PAPPA | |
Polyclonal | |
Unconjugated | |
PAPPA | |
8430414N03Rik; ASBABP2; aspecific BCL2 ARE-binding protein 2; differentially placenta 1 expressed protein; DIPLA1; IGFBP-4 ase; IGFBP-4ase; IGF-dependent IGFBP-4 protease; insulin-like growth factor-dependent IGF binding protein-4 protease; insulin-like growth factor-dependent IGF-binding protein 4 protease; PAG1; PAPA; PAPP A; PAPPA; PAPP-A; PAPPA1; Pappalysin; pappalysin 1; pappalysin-1; pregnacy-associated plasma protein A; pregnancy-associated plasma protein A; pregnancy-associated plasma protein A, pappalysin 1; RP11-45A16.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5069 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q13219 | |
PAPPA | |
E.coli-derived human PAPP A recombinant protein (Position: R95-Q388). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction