Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pappalysin-1/PAPP-A Antibody (10E1cc), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1208346
Description
Pappalysin-1/PAPP-A Monoclonal specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, Immunoprecipitation, Sandwich ELISA.Specifications
Pappalysin-1/PAPP-A | |
Monoclonal | |
Unconjugated | |
ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human PAPP-A antigen purified from placental blood (heterotetrameric complex, consisting of PAPP-A subunits and proMBP subunits). | |
Store at 4C. Do not freeze. | |
IgG2b |
Western Blot, Immunoprecipitation, ELISA | |
10E1cc | |
Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500, Sandwich ELISA 1:100-1:2000 | |
PAPPA | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified human PAPP-A/proMBP (heterotetrameric PAPP-A, htPAPP-A) complex. | |
0.2 mg | |
Cancer | |
5069 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction