Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pappalysin-1/PAPP-A Antibody (9), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105116V
Description
Pappalysin-1/PAPP-A Monoclonal specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Pappalysin-1/PAPP-A | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PAPPA | |
| Purified human PAPP-A | |
| 0.1 mL | |
| Cancer | |
| 5069 | |
| Human, Non-species specific | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 9 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specific for native human PAPP-A. Heat and SDS denatured PAPP-A is also recognized, but not acid treated PAPP-A. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction