Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pappalysin-1/PAPP-A Antibody (PAPP A 1-1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221614AF594
Description
Pappalysin-1/PAPP-A Monoclonal specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pappalysin-1/PAPP-A | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PAPPA | |
Purified human PAPP-A adsorbed onto aluminum hydroxide gel. | |
0.1 mL | |
Primary | |
Specific for native human PAPP-A. | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PAPP A 1-1 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1 | |
Mouse | |
Protein A or G purified | |
Cancer | |
5069.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction