Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAPSS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112226 Supplier Invitrogen™ Supplier No. PA5112226
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAPSS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

Three-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) is the sulfate donor cosubstrate for all sulfotransferase (SULT) enzymes (Xu et al., 2000 ). SULTs catalyze the sulfate conjugation of many endogenous and exogenous compounds, including drugs and other xenobiotics. In humans, PAPS is synthesized from adenosine 5-prime triphosphate (ATP) and inorganic sulfate by 2 isoforms, PAPSS1 and PAPSS2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPSS1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene PAPSS1
Gene Accession No. O43252
Gene Alias 3'-phosphoadenosine 5'-phosphosulfate synthase 1; 3'-phosphoadenosine-5'-phosphosulfate synthase; 3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 1; Adenosine-5'-phosphosulfate 3'-phosphotransferase; Adenylylsulfate 3'-phosphotransferase; Adenylyl-sulfate kinase; AI325286; APS kinase; Asapk; ATPSK1; ATP-sulfurylase; bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 1; PAPS synthase 1; PAPS Synthetase 1; PAPSS; PAPSS 1; PAPSS1; SAT; SK 1; SK1; Sulfate adenylate transferase; Sulfate adenylyltransferase; sulfurylase kinase 1
Gene Symbols PAPSS1
Host Species Rabbit
Immunogen E. coli-derived Human PAPSS1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9061
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.