Learn More
Invitrogen™ PAPSS2 Monoclonal Antibody (OTI2E7)

Description
PAPSS2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PAPSS2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2E7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PAPSS2 |
| Gene Accession No. | O95340 |
| Gene Alias | 1810018P12Rik; 3'-phosphoadenosine 5'-phosphosulfate synthase 2; 3'-phosphoadenosine-5'-phosphosulfate synthase; 3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 2; adenosine 5'-phosphosulfate kinase; Adenosine-5'-phosphosulfate 3'-phosphotransferase; Adenylylsulfate 3'-phosphotransferase; Adenylyl-sulfate kinase; AI159688; APS kinase; ATP sulfurylase/adenosine 5'-phosphosulfate kinase; ATP sulfurylase/APS kinase 2; ATPSK2; AtpsU2; ATP-sulfurylase; BCYM4; Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2; bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2; bm; brachymorphic; PAPS synthase 2; PAPS synthetase 2; PAPS synthethase 2; PAPSS 2; PAPSS2; SAT; SK 2; SK2; Sulfate adenylate transferase; Sulfate adenylyltransferase; sulfurylase kinase 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.