Learn More
PAPSS2 Mouse anti-Human, Clone: OTI5H3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807110

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Sulfation is a common modification of endogenous (lipids, proteins, and carbohydrates) and exogenous (xenobiotics and drugs) compounds. In mammals, the sulfate source is 3′-phosphoadenosine 5′-phosphosulfate (PAPS), created from ATP and inorganic sulfate. Two different tissue isoforms encoded by different genes synthesize PAPS. This gene encodes one of the two PAPS synthetases. Defects in this gene cause the Pakistani type of spondyloepimetaphyseal dysplasia. Two alternatively spliced transcript variants that encode different isoforms have been described for this gene.Specifications
PAPSS2 | |
Monoclonal | |
Unconjugated | |
PAPSS2 | |
ATPSK2, BCYM4, SK2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9060 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95340 | |
PAPSS2 | |
Human recombit protein fragment corresponding to amino acids 1-247 of human PAPSS2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.