Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAPSS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140275 Supplier Invitrogen™ Supplier No. PA5140275
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DTLLDGMALP DGVINMSIPI VLPVSAEDKT R Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 81%.

Sulfation is a common modification of endogenous (lipids, proteins, and carbohydrates) and exogenous (xenobiotics and drugs) compounds. In mammals, the sulfate source is 3'-phosphoadenosine 5'-phosphosulfate (PAPS), created from ATP and inorganic sulfate. Two different tissue isoforms encoded by different genes synthesize PAPS. This gene encodes one of the two PAPS synthetases. Defects in this gene cause the Pakistani type of spondyloepimetaphyseal dysplasia. Two alternatively spliced transcript variants that encode different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPSS2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAPSS2
Gene Accession No. O95340
Gene Alias 1810018P12Rik; 3'-phosphoadenosine 5'-phosphosulfate synthase 2; 3'-phosphoadenosine-5'-phosphosulfate synthase; 3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 2; adenosine 5'-phosphosulfate kinase; Adenosine-5'-phosphosulfate 3'-phosphotransferase; Adenylylsulfate 3'-phosphotransferase; Adenylyl-sulfate kinase; AI159688; APS kinase; ATP sulfurylase/adenosine 5'-phosphosulfate kinase; ATP sulfurylase/APS kinase 2; ATPSK2; AtpsU2; ATP-sulfurylase; BCYM4; Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2; bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2; bm; brachymorphic; PAPS synthase 2; PAPS synthetase 2; PAPS synthethase 2; PAPSS 2; PAPSS2; SAT; SK 2; SK2; Sulfate adenylate transferase; Sulfate adenylyltransferase; sulfurylase kinase 2
Gene Symbols PAPSS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PAPSS2. Recombinant protein control fragment (Product #RP-109467).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9060
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.