Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAQR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524654
Description
This antibody is predicted to react with mouse based on sequence homology.
PAQR3 is function as a spatial regulator of RAF1 kinase by sequestrating it to the Golgi.
Specifications
| PAQR3 | |
| Polyclonal | |
| Unconjugated | |
| PAQR3 | |
| PAQR3; Progestin and adipoQ receptor family member 3; progestin and adipoQ receptor family member III; Raf kinase trapping to Golgi; RKTG | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 152559 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6TCH7 | |
| PAQR3 | |
| KLH conjugated synthetic peptide between 242-270 amino acids from the C-terminal region of human PAQR3. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction