Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAQR4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569886
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
PAQR4 gene ontology annotations for this gene involves signaling receptor activity.
Specifications
PAQR4 | |
Polyclonal | |
Unconjugated | |
PAQR4 | |
1500004C10Rik; LOW QUALITY PROTEIN: progestin and adipoQ receptor family member 4; MNCb-0914; Paqr4; Progestin and adipoQ receptor family member 4; progestin and adipoQ receptor family member IV | |
Rabbit | |
Affinity Chromatography | |
RUO | |
302967 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
PAQR4 | |
synthetic peptide directed towards the middle region of human PAQR4. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction