Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAQR6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567298
Description
Immunogen sequence: YIGEGTPGPAR EEAGADAFPE HRMNWATATS YSTSVQCWAP TSSWRQCWLI WDHAEPGWPH RNLPWAWQAQ WPHWSWLQL Highest antigen sequence identity to the following orthologs - mouse 26%, rat 26%.
Plasma membrane progesterone (P4) receptor coupled to G proteins (PubMed:23763432, PubMed:23161870). Seems to act through a G(s) mediated pathway (PubMed:23161870). Involved in neurosteroid inhibition of apoptosis (PubMed:23161870). May be involved in regulating rapid P4 signaling in the nervous system (PubMed:23763432). Also binds dehydroepiandrosterone (DHEA), pregnanolone, pregnenolone and allopregnanolone (PubMed:23763432, PubMed:23161870). [UniProt].
Specifications
PAQR6 | |
Polyclonal | |
Unconjugated | |
PAQR6 | |
1500001B10Rik; FLJ22672; LOW QUALITY PROTEIN: membrane progestin receptor delta; LOW QUALITY PROTEIN: progestin and adipoQ receptor family member 6; Membrane progesterone P4 receptor delta; Membrane progesterone receptor delta; Membrane progestin receptor delta; MGC137682 protein; mPR delta; Paqr6; PRdelta; Progesterone and adipoQ receptor family member 6; Progestin and adipoQ receptor family member 6; progestin and adipoQ receptor family member VI; progestin and adipoQ receptor family member VI variant 3; progestin and adipoQ receptor family member VI variant 4; progestin and adipoQ receptor family member VI variant 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6TCH4 | |
PAQR6 | |
Recombinant Human PAQR6. Recombinant protein control fragment (Product #RP-105580). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction