Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAQR6 Polyclonal Antibody

Catalog No. PIPA532033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532033 Supplier Invitrogen™ Supplier No. PA532033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain. Predicted reactivity: Mouse (91%), Rat (92%), Pig (92%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plasma membrane progesterone (P4) receptor coupled to G proteins (PubMed:23763432, PubMed:23161870). Seems to act through a G(s) mediated pathway (PubMed:23161870). Involved in neurosteroid inhibition of apoptosis (PubMed:23161870). May be involved in regulating rapid P4 signaling in the nervous system (PubMed:23763432). Also binds dehydroepiandrosterone (DHEA), pregnanolone, pregnenolone and allopregnanolone (PubMed:23763432, PubMed:23161870). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAQR6
Applications Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PAQR6
Gene Accession No. Q6TCG5, Q6TCH4
Gene Alias 1500001B10Rik; FLJ22672; LOW QUALITY PROTEIN: membrane progestin receptor delta; LOW QUALITY PROTEIN: progestin and adipoQ receptor family member 6; Membrane progesterone P4 receptor delta; Membrane progesterone receptor delta; Membrane progestin receptor delta; MGC137682 protein; mPR delta; Paqr6; PRdelta; Progesterone and adipoQ receptor family member 6; Progestin and adipoQ receptor family member 6; progestin and adipoQ receptor family member VI; progestin and adipoQ receptor family member VI variant 3; progestin and adipoQ receptor family member VI variant 4; progestin and adipoQ receptor family member VI variant 5
Gene Symbols PAQR6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PAQR6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68957, 79957
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.