Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAR/pADPr Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. 4335MC100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-35M-C100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 43-35M-C100 Supplier R&D Systems Supplier No. 4335MC100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 22 publications

PAR/pADPr Monoclonal specifically detects PAR/pADPr in Multi-Species samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunoaffinity Purification, Intracellular Staining by Flow Cytometry.

Specifications

Antigen PAR/pADPr
Applications Western Blot, ELISA, Immunoprecipitation, Affinity Purification, Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, ELISA, Immunoprecipitation, Immunoaffinity Purification, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Gene Alias PAR, Poly(ADP-ribose)
Quantity 100 μL
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.