Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519102
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519102 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519102 Supplier Invitrogen™ Supplier No. PA519102
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

Thrombin is a serine protease that is involved in platelet aggregation and blood coagulation. It is cleaved from its precursor, prothrombin, and converts fibrinogen to fibrin in the final step of the clotting cascade. Thrombin mediates its regulatory effects by activating cell surface G-protein coupled receptors which include PAR-1, PAR-2 and PAR3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene F2R
Gene Accession No. P25116
Gene Alias AI482343; C HTR; Cf2r; coagulation factor II (thrombin) receptor; coagulation factor II receptor; coagulation factor II thrombin receptor; F2R; HTR; PAR1; PAR-1; protease activated receptor 1; protease-activated receptor 1; proteinase-activated receptor 1; Thrombin receptor; ThrR; TR; TRGPC
Gene Symbols F2R
Host Species Goat
Immunogen Synthetic peptide sequence (TRARRPESKATNAT) corresponding to the internal amino acids of F2R (aa 24-38).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2149
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.