Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAR1/Thrombin Receptor Antibody (731115), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3855B
Description
PAR1/Thrombin Receptor Monoclonal antibody specifically detects PAR1/Thrombin Receptor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
PAR1/Thrombin Receptor | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
731115 | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, TRproteinase-activated receptor 1 | |
E. coli-derived recombinant human PAR1, Arg27-Thr102, Ser375-Thr425, Accession # P25116 | |
0.1 mL | |
Apoptosis, Cancer, GPCR | |
2149 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction