Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAR1/Thrombin Receptor Antibody (N2-11), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP171770FR
Description
PAR1/Thrombin Receptor Monoclonal specifically detects PAR1/Thrombin Receptor in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
PAR1/Thrombin Receptor | |
Monoclonal | |
DyLight 680 | |
P25116 | |
F2R | |
The oligopeptide CNATLDPRSFLL from human Thrombin Receptor. [Swiss-Prot# P25116] | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
N2-11 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, TRproteinase-activated receptor 1 | |
Mouse | |
52 kDa | |
0.1 mL | |
Apoptosis, Cancer, GPCR | |
2149 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction