Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAR1/Thrombin Receptor Antibody (N2-11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 7 publications

$206.50 - $499.50

Specifications

Antigen PAR1/Thrombin Receptor
Clone N2-11
Concentration 1 mg/mL
Dilution Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100. Use reported in scientific literature (PMID 35055169), Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 31885791), Immunofluorescence
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP171770SS
SDP
View Documents
Novus Biologicals
NBP171770SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP171770
SDP
View Documents
Novus Biologicals
NBP171770
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

PAR1/Thrombin Receptor Monoclonal specifically detects PAR1/Thrombin Receptor in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

PAR1/Thrombin Receptor
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Mouse
Apoptosis, Cancer, GPCR
P25116, P30558
2149
The oligopeptide CNATLDPRSFLL from human Thrombin Receptor. [Swiss-Prot# P25116]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
N2-11
Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100. Use reported in scientific literature (PMID 35055169), Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 31885791), Immunofluorescence
Monoclonal
Purified
RUO
Human, Mouse
CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, TRproteinase-activated receptor 1
F2R
IgG1
Protein G purified
52 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.