Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAR1/Thrombin Receptor Antibody (N2-11) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280904
Description
PAR1/Thrombin Receptor Monoclonal specifically detects PAR1/Thrombin Receptor in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PAR1/Thrombin Receptor | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1:250 - 1:1000, ELISA : 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence : 1:50, Immunohistochemistry-Paraffin : 1:100 | |
CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, TRproteinase-activated receptor 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
N2-11 | |
Unconjugated | |
Tris-Glycine, 0.15 M NaCl | |
F2R | |
The oligopeptide CNATLDPRSFLL from human Thrombin Receptor. [Swiss-Prot# P25116] | |
0.1 mL | |
Apoptosis, Cancer, GPCR | |
2149 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction