Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAR2 Antibody (344222), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3949MFV610
Description
PAR2 Monoclonal antibody specifically detects PAR2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| PAR2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 2150 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 344222 | |
| Flow Cytometry, CyTOF-ready | |
| coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1 | |
| NS0 mouse myeloma cell line transfected with human PAR2, Accession # P55085 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction