Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAR2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP144306
Description
PAR2 Polyclonal specifically detects PAR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PAR2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.1% Sodium Azide | |
F2RL1 | |
Synthetic 18 amino acid peptide from 2nd extracellular domain of human PAR2. | |
0.05 mL | |
GPCR, Signal Transduction | |
2150 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 | |
coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This product is specific for Proteinase-Activated 2 (F2RL1). Unless other species reactivities are specified there is no testing data currently available. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction