Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAR2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2112500.05MG
Description
PAR2 Polyclonal specifically detects PAR2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
PAR2 | |
Polyclonal | |
Unconjugated | |
Antibody stabilization buffer with no preservative | |
F2RL1 | |
Synthetic peptide taken within amino acid region 300-370 of mouse PAR2. | |
Primary | |
This antibody detects a band of approximately 50kDa PAR2 protein in samples and a 52kDa protein in rat hippocampal samples. The antibody also labels a faint band of 110kDa in hippocampal membranes, probably represents either an uncharacterized variant of PAR2 or a non-specific protein. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.50 mg/mL | |
Western Blot 1:250, ELISA 1:1500-1:20000, Immunohistochemistry 1:100, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100 | |
coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1 | |
Rabbit | |
0.05 mg | |
2150 | |
Human, Mouse, Rat, Monkey | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction