Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533529
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533529 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533529 Supplier Invitrogen™ Supplier No. PA533529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset (100%) Monkey, Bovine, Panda (95%) Gibbon, Dog, Elephant, Pig (89%) Rat, Hamster (84%).

This polyclonal antibody was raised in Rabbit against F2RL2 / PAR3. This antibody is unconjugated. This antibody is useful for testing in the following assays: ICC, IHC-P. The specificity of this antibody has been described as follows: Human F2RL2. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene F2RL2
Gene Accession No. O00254
Gene Alias coagulation factor II (thrombin) receptor-like 2; coagulation factor II (thrombin) receptor-like 2 precursor; coagulation factor II receptor-like 2; Coagulation factor II receptor-like 2 (protease-actovated receptor 3); coagulation factor II thrombin receptor like 2; F2RL2; PAR3; PAR-3; protease-activated receptor 3; proteinase-activated receptor 3; proteinase-activated receptor-3; thrombin receptor-like 2
Gene Symbols F2RL2
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from internal region of human F2RL2.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.