Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208155
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8155 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8155 Supplier Fabgennix Inc Supplier No. PAR3301AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAR3 Polyclonal antibody specifically detects PAR3 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen PAR3
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene F2RL2
Gene Accession No. O00254, O08675, Q920E1
Gene Alias coagulation factor II (thrombin) receptor-like 2; coagulation factor II (thrombin) receptor-like 2 precursor; coagulation factor II receptor-like 2; Coagulation factor II receptor-like 2 (protease-actovated receptor 3); coagulation factor II thrombin receptor like 2; F2RL2; PAR3; PAR-3; protease-activated receptor 3; proteinase-activated receptor 3; proteinase-activated receptor-3; thrombin receptor-like 2
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1-100 on mouse Proteinase-activated receptor 3 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14064, 2151, 29636
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.