Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAR4 Monoclonal Antibody (3G9H7,4H12E9)
GREENER_CHOICE

Catalog No. PIMA515408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515408 Supplier Invitrogen™ Supplier No. MA515408
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15408 targets PAR4 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15408 immunogen is purified recombinant fragment of PAR4(aa1-330) expressed in E. Coli..

This gene encodes a tumor suppressor protein that selectively induces apoptosis in cancer cells through intracellular and extracellular mechanisms. The intracellular mechanism involves the inhibition of pro-survival pathways and the activation of Fas-mediated apoptosis, while the extracellular mechanism involves the binding of a secreted form of this protein to glucose regulated protein 78 (GRP78) on the cell surface, which leads to activation of the extrinsic apoptotic pathway. This gene is located on the unstable human chromosomal 12q21 region and is often deleted or mutated different tumors. The encoded protein also plays an important role in the progression of age-related diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAR4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3G9H7,4H12E9, 4H12E9
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene Pawr
Gene Accession No. Q96IZ0
Gene Alias 2310001G03Rik; coagulation factor II (thrombin) receptor-like 3; Coagulation factor II receptor-like 3; F2R like thrombin/trypsin receptor 3; F2RL3; PAR4; Par-4; par-4 induced by effectors of apoptosis; Pawr; PRKC apoptosis WT1 regulator protein; PRKC, apoptosis, WT1, regulator; pro-apoptotic WT1 regulator; prostate apoptosis response 4; prostate apoptosis response 4 protein; Prostate apoptosis response protein 4; prostate apoptosis response protein PAR-4; prostate apoptosis response-4; protease-activated receptor-4; proteinase-activated receptor 4; Thrombin receptor-like 3; transcriptional repressor PAR4; transcriptional repressor Par-4-like protein PAWR; WT1-interacting protein
Gene Symbols Pawr
Host Species Mouse
Immunogen Purified recombinant fragment of PAR4(aa1-330) expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.