Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PAR4, Mouse, Unlabeled, Clone: 14H6,

Catalog No. MABS1299MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1299MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1299MI Supplier MilliporeSigma™ Supplier No. MABS1299
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Proteinase-activated receptor 4 (UniProt Q96RI0; also known as Coagulation factor II receptor-like 3, PAR-4, Thrombin receptor-like 3) is encoded by the F2RL3 (also known as PAR4) gene (Gene ID 9002) in human. Protease-activated receptors (PARs) constitute a unique family of seven-transmembrane, G-protein-coupled receptors (GPCRs) activated by proteolytic cleavage of their N-terminal propeptide sequence. Once cleaved off, the N-terminal propeptide fragment functions as a ligand and activates the receptor by binding the second extracellular loop. The four PAR family members (PAR-1 to PAR-4) are widely expressed and activated by multiple proteases, and utilize different types of G-proteins (Gi, Gq, and G12/13) for signal transdution depending on the activating protease and cellular context. PAR-4 is expressed on platelets and exhibits a low-affinity for thrombin. However, PAR-4 is able to form hetero-oligomers with both PAR-1 and the ADP receptor P2Y12 to mediate thrombin- and ADP-initiated signaling. PAR-4 cleavage is significantly enhanced through hetero-oligomerization with PAR-1, and PAR-4 interaction with P2Y12 is directly linked to arrestin-2 recruitment and AKT signaling. PAR-4 is a 7-transmembrane (a.a. 83-103, 109-129, 152-172, 192-213, 248-268, 284-304, 320-343) GPCR activated by thrombin cleavage between R47 and G48, having 3 extracellular loops and 3 intracellular loops between the extracellular N-terminal end (a.a. 48-82) the cytoplasmic C-terminal tail (a.a. 344-385).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAR4
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 14H6
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene F2RL3, PAR4
Gene Accession No. Q96RI0
Host Species Mouse
Immunogen MBP-conjugated recombinant human PAR4 N-terminal fragment including the propeptide sequence.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_003941
Test Specificity Clone 14H6 recognizes an epitope near (C-terminal) to the thrombin-cleavage site. Unlike clone 5F10 (Mfr. No. MABS1298), clone 14H6 does not protect cell surface PAR4 against thrombin cleavage (Mumaw, M.M., et al. (2015). Thromb. Res. 135(6):1165-1171). Clone 14H6 recognizes prepro- and pro-, but not proteolytically activated, human PAR4. A structual change at the cleavage site following thrombin cleavage is believed to prevent the detection of the activated PAR-4 by clone 5F10.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.