Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1755
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1755 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1755 Supplier Invitrogen™ Supplier No. PA1755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-755 detects prostate apoptosis response protein (Par-4) from human and rat tissues. PA1-755 has been sucessfully used in Western blot procedures. By Western blot, this antibody detects an ~40 kDa protein representing Par-4 in AT-3 cell extract. In Western blots of rat tissue extracts, including testes, spleen, ovary, lung, and skeletal muscle, ~40 kDa and ~38 kDa proteins representing Par-4 are detected. In Western blots of T47D cell extracts, an ~40 kDa protein representing Par-4 and a smaller ~20 kDa unknown protein is detected. PA1-755 immunizing peptide corresponds to amino acids residues 322-340 from human Par-4. PA1-755 immunizing peptide (Cat. # PEP-060) is available for use in neutralization and control experiments.

This gene encodes a tumor suppressor protein that selectively induces apoptosis in cancer cells through intracellular and extracellular mechanisms. The intracellular mechanism involves the inhibition of pro-survival pathways and the activation of Fas-mediated apoptosis, while the extracellular mechanism involves the binding of a secreted form of this protein to glucose regulated protein 78 (GRP78) on the cell surface, which leads to activation of the extrinsic apoptotic pathway. This gene is located on the unstable human chromosomal 12q21 region and is often deleted or mutated different tumors. The encoded protein also plays an important role in the progression of age-related diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAR4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Pawr
Gene Accession No. Q62627, Q96IZ0
Gene Alias 2310001G03Rik; coagulation factor II (thrombin) receptor-like 3; Coagulation factor II receptor-like 3; F2R like thrombin/trypsin receptor 3; F2RL3; PAR4; Par-4; par-4 induced by effectors of apoptosis; Pawr; PRKC apoptosis WT1 regulator protein; PRKC, apoptosis, WT1, regulator; pro-apoptotic WT1 regulator; prostate apoptosis response 4; prostate apoptosis response 4 protein; Prostate apoptosis response protein 4; prostate apoptosis response protein PAR-4; prostate apoptosis response-4; protease-activated receptor-4; proteinase-activated receptor 4; Thrombin receptor-like 3; transcriptional repressor PAR4; transcriptional repressor Par-4-like protein PAWR; WT1-interacting protein
Gene Symbols Pawr
Host Species Rabbit
Immunogen Synthetic Peptide: C E(322) Q L K Q E N K T L L K V V G Q L T R(340).
Purification Method Antigen affinity chromatography
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5074, 64513
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.