Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569202 Supplier Invitrogen™ Supplier No. PA569202
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 87%; Yeast: 85%.

This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 domain and a semi-Cdc42/Rac interactive binding domain. This cell membrane protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex. The protein also has a role in the epithelial-to-mesenchymal transition that characterizes the invasive phenotype associated with metastatic carcinomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAR6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PARD6A
Gene Accession No. Q9Z101
Gene Alias 0710008C04Rik; 2610010A15Rik; PAR6; Par-6; par-6 (partitioning defective 6,) homolog alpha; PAR-6 alpha; par-6 family cell polarity regulator alpha; par-6 partitioning defective 6 homolog alpha; PAR6A; PAR-6A; PAR6alpha; PAR6C; Pard6a; Partitioning defective 6 homolog alpha; partitioning defective protein 6A; partitioning defective-6 homolog alpha; partitioning-defective protein 6; similar to Partitioning defective-6 homolog alpha (PAR-6 alpha) (PAR-6A) (PAR-6) (PAR6C) (Tax interaction protein 40) (TIP-40); tax interaction protein 40; TAX40; Tax-interacting protein 40; Tip-40
Gene Symbols PARD6A
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse PAR6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56513
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.