Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-574 Supplier Invitrogen™ Supplier No. PA3911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA3-911detects Par-6 from human and mouse samples. PA3-911has been successfully used in Western blot and ICC/IF procedures. This antibody is not recommended for U87-MG cells in Western blot applications. The PA3-911 immunogen is a combination of two synthetic peptides coupled to KLH.

This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 domain and a semi-Cdc42/Rac interactive binding domain. This cell membrane protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex. The protein also has a role in the epithelial-to-mesenchymal transition that characterizes the invasive phenotype associated with metastatic carcinomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAR6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole Serum with 0.05% sodium azide
Gene PARD6A
Gene Accession No. Q9NPB6
Gene Alias 0710008C04Rik; 2610010A15Rik; PAR6; Par-6; par-6 (partitioning defective 6,) homolog alpha; PAR-6 alpha; par-6 family cell polarity regulator alpha; par-6 partitioning defective 6 homolog alpha; PAR6A; PAR-6A; PAR6alpha; PAR6C; Pard6a; Partitioning defective 6 homolog alpha; partitioning defective protein 6A; partitioning defective-6 homolog alpha; partitioning-defective protein 6; similar to Partitioning defective-6 homolog alpha (PAR-6 alpha) (PAR-6A) (PAR-6) (PAR6C) (Tax interaction protein 40) (TIP-40); tax interaction protein 40; TAX40; Tax-interacting protein 40; Tip-40
Gene Symbols PARD6A
Host Species Rabbit
Immunogen Combination of two synthetic peptides coupled to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50855
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.