Learn More
Thermo Scientific™ para-JEM™ Reagent Kit
Click to view available options
Packaging Type:
250 Tests/Kit
Description
Achieve faster detection of M. avium ssp. paratuberculosis with the fully automated Thermo Scientific™ VersaTREK™ System and Thermo Scientific™ para-JEM™ Reagent Kit. Kit includes para-JEM Broth, para-JEM Growth Supplement, para-JEM Antibiotic Supplement and connectors for 250 tests on the VersaTREK System.
Solid medium culture used to be the standard procedure for detecting M. avium ssp. paratuberculosis (MAP), the causative agent of Johne's Disease, in ruminant fecal samples before para-JEM reagents. Due to the bacterium's long generation time, it can take up to 12 weeks of incubation on solid medium before the bacteria are detected. The para-JEM System is a unique, liquid culture method that combines a full line of reagents with the powerful Thermo Scientific™ VersaTREK™ Automated Microbial Detection System for faster, automated detection of MAP.
The para-JEM System includes a number of unique features:
Not all products are available for sale in all territories. Please inquire.
The para-JEM System includes a number of unique features:
- Detects faster—able to recover MAP in as little as 2 to 3 weeks, much earlier than conventional methods*
- Provides increased sensitivity over conventional methods*
- Provides semi-quantitation for low, medium and high shedders based on time-to-detection*
- Reduces labor with fewer false positives, false negatives and contaminants*
- Improves workflow with most tests concluded at six weeks for high, medium and many low shedders*
Not all products are available for sale in all territories. Please inquire.
Specifications
Specifications
| Description | para-JEM Reagent Kit |
| Product Type | para-JAM Microbial Detection |
| Test Type | Microbial Detection |
| No. per Pack | 250 Tests/Kit |
| Packaging Type | 250 Tests/Kit |
| Specimen Type | Ruminant Fecal Samples |
| Species | M. avium ssp. paratuberculosis |