Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetraphenylboron sodium, ACS reagent
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >300.0°C |
| Color | Colorless to White |
| Physical Form | Crystals or Granular Powder |
| Quantity | 500 g |
| Linear Formula | (C6H5)4BNa |
| Merck Index | 15,8818 |
| Solubility Information | Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in acetone, slightly soluble in ether and chloroform, practically insoluble in petroleum ether |
| Formula Weight | 342.21 |
| Grade | ACS Reagent |
| Infrared Spectrum | Authentic |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Call a POISON CENTER or doctor/physician if you feel unwell. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective cloth |
| EINECSNumber | 205-605-5 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction