Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heathrow Scientific Parafilm™ Sterile Strips
Click to view available options
Length (Metric):
15.2 cm
20.3 cm
Description
- Ideal for protecting samples and preventing contamination or evaporation perfect for cleanroom environments and strict sanitation protocols
- These pre-cut strips are sterilized using a Vapor Hydrogen Peroxide process, adding an extra layer of protection to Parafilm natural waterproof and moisture-resistant properties
- Parafilm™, semi-transparent thermoplastic film designed for reliable sealing in laboratory, medical, and industrial environments
- Engineered from a unique blend of waxes and polyolefins, Parafilm™streches up to 200 % original size, conforming tightly to irregular surfaces such as test tubes, beakers, petri dishes, and flasks
- Each strip comes individually packaged in a sealed, lot-traceable pouch featuring a color sterilization indicator ensuring both safety and convenience
Specifications
Specifications
Length (English) | 6 in. |
CAS | 67-56-1 |
Product Type | Parafilm Wrap |
Length (Metric) | 15.2 cm |
Material | Thermoplastic |
Thickness | 5 mil |
Linear Formula | C11H12D5NO · HCl |
UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
Concentration | 100 μg/mL in methanol (as free base) |
Formula Weight | 220.75 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction