Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paraformaldehyde, 90%, pure

Catalog No. p-4510509
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bucket
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 30525-89-4 | CH2O | 30.026 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point 120.0°C to 170.0°C
Color White to off-white
pH 3.5 to 5 (10% suspension)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 89.5 to 93%
Packaging Plastic bottle
Linear Formula (CH2O)n
Quantity 1 kg
Beilstein 01,566
Merck Index 15,7129
Solubility Information Solubility in water: sparingly soluble. Other solubilities: the higher polymers are insoluble in water, insoluble in alcohol and ether
Percent Purity 90%
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Paraformaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
Harmful if inhaled.<br
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.