Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Parainfluenza Virus Type 1 Monoclonal Antibody (D57D)
GREENER_CHOICE

Catalog No. MA17074
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17074 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17074 Supplier Invitrogen™ Supplier No. MA17074
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7074 detects the Parainfluenza virus type 1. MA1-7074 has been successfully used in immunofluorescence procedures. The MA1-7074 immunogen is Parainfluenza virus type 1 lysate.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Parainfluenza Virus Type 1
Applications Immunocytochemistry
Classification Monoclonal
Clone D57D
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias canine parainfluenza virus; hPIV-1; PIV-1
Host Species Mouse
Immunogen Parainfluenza virus type 1 lysate.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.