Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Parainfluenza Virus Type 1/3 Monoclonal Antibody (D67F)
GREENER_CHOICE

Catalog No. PIMA17311
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17311 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17311 Supplier Invitrogen™ Supplier No. MA17311
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7311 antibody reacts with Parainfluenza virus type 1 in viral and infected tissue samples. MA1-7311 has been successfully used in ELISA applications. The MA1-7311 antibody was raised against Parainfluenza Type 1 antigen.

Parainfluenza Virus Type 1/3 is a respiratory virus that primarily affects young children and infants. It is highly contagious and can cause symptoms such as coughing, fever, and difficulty breathing. The virus can lead to more severe respiratory illnesses, such as croup and bronchiolitis, and can be particularly dangerous for individuals with weakened immune systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parainfluenza Virus Type 1/3
Applications ELISA
Classification Monoclonal
Clone D67F
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias canine parainfluenza virus; hPIV-1; PIV-1
Host Species Mouse
Immunogen Parainfluenza Type 1 antigen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.