Learn More
Invitrogen™ Parainfluenza Virus Type 1 Polyclonal Antibody, Biotin

Description
The PA1-73043 antibody reacts with Parainfluenza type 1. PA1-73043 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73043 antibody was raised against Parainfluenza cantell strain.
Specifications
Specifications
| Antigen | Parainfluenza Virus Type 1 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | canine parainfluenza virus; hPIV-1; PIV-1 |
| Host Species | Goat |
| Immunogen | Parainfluenza cantell strain. |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.