Learn More
Invitrogen™ Parainfluenza Virus Type 2/3 Polyclonal Antibody, Biotin
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA173041
Description
The PA1-73041 antibody reacts with Parainfluenza types 2 & 3. PA1-73041 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73041 antibody was raised against Parainfluenza type 3 isolated from human serum. Store 2-8°C for up to 6 months. For long term storage aliquot and freeze.

Specifications
Parainfluenza Virus Type 2/3 | |
Polyclonal | |
Biotin | |
canine parainfluenza virus; hPIV-2; human parainfluenza virus type 2; PIV-2; PIV-3 | |
Parainfluenza type 3 isolated from human serum. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
IgG fraction | |
RUO | |
Virus | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.