Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Parainfluenza Virus Type 3 Monoclonal Antibody (F96H)
GREENER_CHOICE

Catalog No. PIMA17317
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17317 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17317 Supplier Invitrogen™ Supplier No. MA17317
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7317 antibody reacts with Parainfluenza virus type 3 in viral and infected tissue samples. MA1-7317 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7317 antibody was raised against Parainfluenza Type 3 antigen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Parainfluenza Virus Type 3
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone F96H
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias canine parainfluenza virus; hPIV-3; PIV-3
Host Species Mouse
Immunogen Parainfluenza Type 3 antigen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.