Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parainfluenza virus type 3 Mouse anti-Virus, Clone: HM180, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13630
Description
Mouse monoclonal antibody raised against parainfluenza virus type 3 (PIV-3).
- High specificity
- High reactivity
Specifications
Parainfluenza virus type 3 | |
Monoclonal | |
Unconjugated | |
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
Mouse | |
1 mg | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Immunofluorescence, Lateral Flow | |
HM180 | |
Mouse monoclonal antibody raised against parainfluenza virus type 3 (PIV-3). | |
In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
Protein A purification | |
RUO | |
Virus | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction