Learn More
Invitrogen™ Parainfluenza Virus Type 3 Polyclonal Antibody

Description
PA1-7235 detects the parainfluenza virus, type 3. This antibody cross-reacts with type 2, and has minimal reactivity with type 1. PA1-7235 has been successfully used in immunofluorescence procedures. The PA1-7235 immunogen is human isolate, type 3. Recommended short term (<6months) storage is liquid at 2-8ºC. For long term, aliquot and freeze.
Specifications
Specifications
| Antigen | Parainfluenza Virus Type 3 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | canine parainfluenza virus; hPIV-3; PIV-3 |
| Host Species | Goat |
| Immunogen | Human isolate, type 3. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.