Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stoelting™ Parallel Rail Stereotaxic Instruments for Large Animals

Designed for large animals, but can be used on almost any laboratory animal when used with Stoelting’s selection of animal species adaptors
$1124.00 - $23320.00
Specifications
For Use With (Application) | Neuroscience |
---|---|
Animal | Large Animal (But can be adapted for any size animal with appropriate adaptor) |
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
10-000-209
![]() |
Stoelting™
51800 |
Parallel Rail Stereotaxic |
Each for $20,630.00
|
|
|||||
10-000-211
![]() |
Stoelting™
51804 |
Manipulator on Rail Clamp, LH |
Each for $5,030.00
|
|
|||||
10-000-212
![]() |
Stoelting™
51806 |
Manipulator on Rail Clamp, RH |
Each for $5,030.00
|
|
|||||
10-000-224
![]() |
Stoelting™
51850 |
Large Parallel Rail Stereotaxic |
Each for $23,320.00
|
|
|||||
10-000-227
![]() |
Stoelting™
51857 |
Rail Clamp |
Each for $1,124.00
|
|
|||||
10-000-228
![]() |
Stoelting™
51858 |
A-P Microdrive, on Rail Clamp |
Each for $4,610.00
|
|
|||||
10-000-229
![]() |
Stoelting™
51871 |
Base Plate, Plain, 17 X 13in |
Each for $1,610.00
|
|
|||||
Description
- Up to six manipulator arms may be positioned on the two parallel rails. Thus, several probes may be independently positioned in one animal.
- Reliable Locks
- Laser Engrave Scales
- Precise, laser engraved graduations allow vernier scales to be aligned to 0.1 mm
- Scales are oriented to enhance readability from common user positions
- Standard manipulator arms have 100 micron resolution
- Digital arms have a resolution of 10 microns
- Accurate
- Versatile - Other species adaptors, probe holders and ear bars manufactured by Stoelting may be used on the 51800 series of stereotaxic instruments allowing for a use with a wide range of animals and procedures
- Works with large range of animals
- AP Microdrive available for fine control of third axis
Specifications
Neuroscience | |
Large Animal (But can be adapted for any size animal with appropriate adaptor) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title