Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Paramyxovirus SV5 Pk Antibody (SV5-Pk1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062264H
Description
Paramyxovirus SV5 Pk Monoclonal specifically detects Paramyxovirus SV5 Pk in Mouse, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Paramyxovirus SV5 Pk | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein G purified | |
RUO | |
This product recognises a small epitope, termed Pk, present on the P/V proteins of the paramyxovirus, SV5. NB100-62264 has been used to detect recombinant proteins, some of which include transmembrane and secreted proteins, which have been tagged with this epitope. Usually, a 14 amino acid tag has been added to the recombinant proteins, although a smaller epitope of 9 amino acids (that as a peptide inhibit the binding of the monoclonal antibody to its native protein) has also been successfully used. The 14 amino acid epitope is; gly lys pro (ile pro asn pro leu leu gly leu asp) ser thr. (9 amino acid epitope is in parenthesis) | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
SV5-Pk1 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
Simian virus 5; SPV5gp2; V protein | |
Mice were infected with the paramyxovirus SV5, Simian-Virus 5 | |
0.1 mL | |
Primary | |
Mouse, Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction