Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Paraquat Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA185762
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA185762 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA185762 Supplier Invitrogen™ Supplier No. PA185762
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Sheep anti Paraquat antibody recognizes the herbicide Paraquat, also known as N,N'-dimethyl-4,4'-bipyridinium dichloride, and shows the following reactivities with related pesticides: Monoquat 5%, Diquat, and Norfamaquat.

Paraquat is the trade name for N, N -dimethyl-4, 4 -bipyridinium dichloride, one of the most widely used herbicides in the world. Paraquat, a viologen, is quick-acting and non-selective, killing green plant tissue on contact. It is redistributed within the plant but does not harm mature bark. Paraquat can protect crops by controlling a wide range of annual and certain perennial weeds that reduce crop yield and quality by competing with the crop for water, nutrients, and light. Pure paraquat is highly toxic to mammals and humans if ingested, potentially leading to acute respiratory distress syndrome (ARDS). There are no specific antidotes. A large majority (93%) of fatalities from paraquat poisoning are cases of intentional self-administration, ie, suicides. [National Center for Biotechnology Information. PubChem Compound Database; CID=15939].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Paraquat
Applications Radioimmune Assays (RIA)
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias C12H14N2+2; Dimethyl viologen; Paraquat dication; Paraquat ion; Starfire
Host Species Sheep
Immunogen Synthetic peptide corresponding to part of the native molecule conjugated to KLH.
Purification Method Protein G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.