Learn More
Epredia™ Lab Vision™ Parathyroid Hormone Receptor (PTH-Receptor) Type 1 Ab-1, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 3D1.1
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: aa 146-169
Immunogen: A 23 amino acid peptide of the hPTH/PTHrP receptor which is not shared by the type II receptor
Molecular Weight: 80 and 160kDa
Positive Control: Breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Parathyroid Hormone Receptor (PTH-Receptor) Type 1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 3D1.1 |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Mouse |
| Immunogen | A 23 amino acid peptide of the hPTH/PTHrP receptor which is not shared by the type II receptor |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.