Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARC Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002300PCP
Description
PARC Polyclonal antibody specifically detects PARC in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| PARC | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| CUL-9, cullin 9, cullin-9, DKFZp686G1042, H7AP1DKFZp686P2024, p53-associated parkin-like cytoplasmic protein, PARCKIAA0708, parkin-like cytoplasmic p53 binding protein, RP3-330M21.2, UbcH7-associated protein 1 | |
| Maps to a region between residues 900 and 950 of human P53-associated parkin-like cytoplasmic protein using the numbering given in TrEMBL entry Q8IWT3 (GeneID 23113). | |
| 0.1 mL | |
| Cancer, DNA Repair, Tumor Suppressors | |
| 23113 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction