Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARD-3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985011
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5011 Supplier Bioss Supplier No. BS2921R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.

Specifications

Antigen PARD-3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Pard3
Gene Accession No. Q8TEW0
Gene Alias AA960621; AI256638; ASBP; ASIP; atypical PKC isotype-specific interacting protein; Atypical PKC isotype-specific-interacting protein; atypical PKC-specific binding protein; Atypical PKC-specific-binding protein; Baz; bazooka; CTCL tumor antigen se2-5; D8Ertd580e; Ephrin-interacting protein; FLJ21015; Par3; PAR-3; par-3 (partitioning defective 3) homolog; par-3 family cell polarity regulator; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3A; PAR3alpha; PAR3-alpha; Pard3; PARD-3; PARD3A; Partitioning defective 3 homolog; partitioning-defective 3 homolog; PHIP; PPP1R118; protein phosphatase 1, regulatory subunit 118; RP11-406D17.1; SE2-5L16; SE2-5LT1; SE2-5T2; three-PDZ containing protein similar to C. elegans PAR3 (partitioning defect)
Gene Symbols Pard3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PARD-3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56288
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.