Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARD3/Par3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
$682.00
Specifications
Antigen | PARD3/Par3 |
---|---|
Concentration | 0.3mg/mL |
Dilution | Western Blot 0.04-0.4 ug/ml, Simple Western 1:1000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation Reported in scientific literature (PMID:34158849)., Immunohistochemistry-Paraffin 1:200-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
PARD3/Par3 Polyclonal specifically detects PARD3/Par3 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
PARD3/Par3 | |
Western Blot 0.04-0.4 ug/ml, Simple Western 1:1000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation Reported in scientific literature (PMID:34158849)., Immunohistochemistry-Paraffin 1:200-1:500 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
56288 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LKGLGDMFRIQAKTREFRERQARERDYAEIQDFHRTFGCDDELMYGGVSSYEGSMALNARPQSPREGHMMDALYAQVKKPRNSKPSPVDSNR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.3mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
FLJ21015, par-3 partitioning defective 3 homolog (C. elegans), PAR3A, PAR3C.elegans) homolog | |
PARD3 | |
IgG | |
Affinity Purified | |
Specificity of human PARD3/Par3 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title